Learn More
Benzoyl fluoride, 97%
CAS: 455-32-3 | C7H5FO | 124.11 g/mol
$76.50 - $76.50
Chemical Identifiers
| CAS | 455-32-3 |
|---|---|
| Molecular Formula | C7H5FO |
| Molecular Weight (g/mol) | 124.11 |
| MDL Number | MFCD00000364 |
| InChI Key | HPMLGNIUXVXALD-UHFFFAOYSA-N |
| Synonym | benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa |
| PubChem CID | 67999 |
| IUPAC Name | benzoyl fluoride |
| SMILES | FC(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC402040050
|
Thermo Scientific Chemicals
402040050 |
5 g |
Each for $76.50
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 455-32-3 | |
| 124.11 | |
| HPMLGNIUXVXALD-UHFFFAOYSA-N | |
| 67999 | |
| FC(=O)C1=CC=CC=C1 |
| C7H5FO | |
| MFCD00000364 | |
| benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
| benzoyl fluoride |
Specifications
| 455-32-3 | |
| 1.1500g/mL | |
| 48°C | |
| 96% min. (GC) | |
| C7H5FO | |
| C6H5COF | |
| 5 g | |
| 1.15 | |
| Solubility in water: hydrolyses. | |
| FC(=O)C1=CC=CC=C1 | |
| 124.11 | |
| 124.11 | |
| Benzoyl fluoride |
| -28.0°C | |
| 159.0°C to 161.0°C | |
| Authentic | |
| Plastic bottle | |
| 1.496 | |
| MFCD00000364 | |
| 09,181 | |
| benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
| HPMLGNIUXVXALD-UHFFFAOYSA-N | |
| benzoyl fluoride | |
| 67999 | |
| 97% |
Safety and Handling
GHS H Statement Flammable liquid and vapor. Causes severe skin burns and eye damage.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
GHS Signal Word: Danger
EINECSNumber : 207-244-9
RUO – Research Use Only