missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl bromide, 97%

Catalog No. AC160950050
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160950050 5 mL Glass bottle
AC160950250 25 mL Glass bottle
AC160951000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160950050 Supplier Thermo Scientific Chemicals Supplier No. 160950050

CAS: 618-32-6 | C7H5BrO | 185.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-32-6
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00000112
InChI Key AQIHMSVIAGNIDM-UHFFFAOYSA-N
Synonym benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide
PubChem CID 12056
IUPAC Name benzoyl bromide
SMILES BrC(=O)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -24.0°C
Color Orange
Density 1.5700g/mL
Boiling Point 218.0°C to 219.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5880 to 1.5900
Linear Formula C6H5COBr
Quantity 5 mL
Beilstein 09,195
Specific Gravity 1.57
Solubility Information Solubility in water: may decompose. Other solubilities: miscible in ethyl ether
Formula Weight 185.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzoyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-544-2

RUO – Research Use Only