missing translation for 'onlineSavingsMsg'
Learn More

Benzoxazole, 97+%

Catalog No. AC105680050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105680050 5 g
AC105680250 25 g
AC105681000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105680050 Supplier Thermo Scientific Chemicals Supplier No. 105680050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 273-53-0 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 273-53-0
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00005765
InChI Key BCMCBBGGLRIHSE-UHFFFAOYSA-N
Synonym benzoxazole, 1-oxa-3-azaindene, 1-oxa-3-aza-1h-indene, benzo d oxazole, benzooxazole, usaf ek-5017, unii-j233y1i55i, benzoxazol, benzoxazoline, benzothiazoie
PubChem CID 9228
ChEBI CHEBI:38814
IUPAC Name 1,3-benzoxazole
SMILES O1C=NC2=CC=CC=C12

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 27°C to 30°C
Color Brown-Orange to Yellow
Boiling Point 182°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 42
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,sulfuric acid
Formula Weight 119.12
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Benzoxazole, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-988-9
RTECSNumber DM4375000
TSCA TSCA

RUO – Research Use Only