missing translation for 'onlineSavingsMsg'
Learn More

Benzopurpurin 4B

Catalog No. AAJ6490122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6490122 100 g
AAJ6490114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6490122 Supplier Thermo Scientific Chemicals Supplier No. J6490122
Only null left

CAS: 992-59-6 | C34H26N6Na2O6S2 | 724.718 g/mol

Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.

Solubility
Soluble in water (20 mg/ml), acetone, ethanol (3 mg/ml), sodium hydroxide, and sulfuric acid.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 992-59-6
Molecular Formula C34H26N6Na2O6S2
Molecular Weight (g/mol) 724.718
MDL Number MFCD00012410
InChI Key SUXCALIDMIIJCK-UHFFFAOYSA-L
Synonym benzopurpurine 4b, benzopurpurin 4b, direct red 2, c.i. direct red 2, amanil purpurine, diphenyl red 4b, hispamin red 4b, direct red dcb, azocard red 4b, diazine red 4b
PubChem CID 13816
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]-2-methylphenyl]diazenyl]naphthalene-1-sulfonate
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=NC3=C(C4=CC=CC=C4C(=C3)S(=O)(=O)[O-])N)C)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N.[Na+].[Na+]

Specifications

Color Red
Quantity 100 g
Merck Index 14,1011
Solubility Information Soluble in water (20mg/ml),acetone,ethanol (3mg/ml),sodium hydroxide,and sulfuric acid.
Formula Weight 724.73
Physical Form Powder
Chemical Name or Material Benzopurpurin 4B
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-594-3
RTECSNumber QK1765000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only