missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzopurpurin 4B
Description
Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.
Solubility
Soluble in water (20 mg/ml), acetone, ethanol (3 mg/ml), sodium hydroxide, and sulfuric acid.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.
Solubility
Soluble in water (20 mg/ml), acetone, ethanol (3 mg/ml), sodium hydroxide, and sulfuric acid.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Color | Red |
| Quantity | 25 g |
| Merck Index | 14,1011 |
| Solubility Information | Soluble in water (20mg/ml),acetone,ethanol (3mg/ml),sodium hydroxide,and sulfuric acid. |
| Formula Weight | 724.73 |
| Physical Form | Powder |
| Chemical Name or Material | Benzopurpurin 4B |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 213-594-3 |
| RTECSNumber | QK1765000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only