missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile 99.0+%, TCI America™

Catalog No. p-7118341
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B008225G 25 g
B0082500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B008225G Supplier TCI America Supplier No. B008225G
Only null left

Chemical Identifiers

CAS 100-47-0
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD00001770
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

Color Colorless
Boiling Point 191°C
Quantity 25 g
UN Number 2224
Formula Weight 103.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzonitrile
EINECSNumber (3)-1796
RTECSNumber DI2450000
TSCA Yes