missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, 99+%, for spectroscopy

Catalog No. p-64055
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167670500 50 mL Glass bottle
AC167672500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167670500 Supplier Thermo Scientific Chemicals Supplier No. 167670500
Only null left

CAS: 100-47-0 | C7H5N | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

Chemical Identifiers

CAS 100-47-0
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.12
MDL Number MFCD00001770
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

CAS Min % 99.0
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 360nm, 0.01 max. at 400nm, 0.03 max. at 335nm, 0.15 max. at 310nm, 0.3 max. at 305nm, 1 max. at 300nm
CAS Max % 100.0
Melting Point -13.0°C
Density 1.0100g/mL
Boiling Point 188.0°C to 191.0°C
Flash Point 71°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290
Linear Formula C6H5CN
Quantity 50 mL
Residue after Evaporation 0.0005% max.
Beilstein 09, 275
Specific Gravity 1.01
Merck Index 15, 1099
Solubility Information Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
Viscosity 1.24 mPa.s (25°C)
Formula Weight 103.12
Percent Purity 99+%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material Benzonitrile, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 202-855-7
RTECSNumber DI2450000
TSCA TSCA

RUO – Research Use Only