missing translation for 'onlineSavingsMsg'
Learn More

Benzoin Zone Refined (number of passes:40) 99.0+%, TCI America™

Catalog No. B0222ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0222ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0222ZONE Supplier TCI America Supplier No. B02221SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004496
InChI Key ISAOCJYIOMOJEB-UHFFFAOYNA-N
Synonym benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone
PubChem CID 8400
ChEBI CHEBI:17682
IUPAC Name 2-hydroxy-1,2-diphenylethan-1-one
SMILES OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C
Color White
Boiling Point 344°C
Quantity ZONE
Formula Weight 212.25
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material Benzoin Zone Refined (number of passes:40)
EINECSNumber (9)-1315
RTECSNumber DI1590000
TSCA Yes