missing translation for 'onlineSavingsMsg'
Learn More

Benzoin Methyl Ether 98.0+%, TCI America™

Catalog No. p-7118720
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B086925G 25 g
B0869500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B086925G Supplier TCI America Supplier No. B086925G
Only null left

Chemical Identifiers

CAS 3524-62-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00008492
InChI Key BQZJOQXSCSZQPS-UHFFFAOYSA-N
Synonym benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
PubChem CID 98097
IUPAC Name 2-methoxy-1,2-diphenylethanone
SMILES COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 189°C
Quantity 25 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoin Methyl Ether
EINECSNumber (3)-1264&(4)-0079
RTECSNumber AM9350000
TSCA Yes