missing translation for 'onlineSavingsMsg'
Learn More

Benzoin Ethyl Ether 99.0+%, TCI America™

Catalog No. B101925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B101925G 25 g
B1019100G 100 g
B1019500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B101925G Supplier TCI America Supplier No. B101925G

Chemical Identifiers

CAS 574-09-4
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.302
MDL Number MFCD00009242
InChI Key KMNCBSZOIQAUFX-UHFFFAOYSA-N
Synonym benzoin ethyl ether, ethanone, 2-ethoxy-1,2-diphenyl, 2-ethoxy-2-phenylacetophenone, 2-ethoxy-1,2-diphenylethan-1-one, 2-ethoxy-1,2-diphenyl-ethanone, 2-ethoxybenzoin, benzoinethylether, ethyl benzoin ether, 2-ethoxy-2-phenyl acetophenone
PubChem CID 101778
IUPAC Name 2-ethoxy-1,2-diphenylethanone
SMILES CCOC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 125°C
Quantity 25 g
UN Number 3077
Formula Weight 240.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoin Ethyl Ether
EINECSNumber (4)-0079
TSCA Yes