missing translation for 'onlineSavingsMsg'
Learn More

Benzoin, 98%

Catalog No. p-64019
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105511000 100 g Plastic bottle
AC105515000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105511000 Supplier Thermo Scientific Chemicals Supplier No. 105511000
Only null left

CAS: 119-53-9 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004496
InChI Key ISAOCJYIOMOJEB-UHFFFAOYNA-N
Synonym benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone
PubChem CID 8400
ChEBI CHEBI:17682
IUPAC Name 2-hydroxy-1,2-diphenylethanone
SMILES OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 133.0°C to 137.0°C
Color White to Yellow
Boiling Point 194.0°C (12.0 mmHg)
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CH(OH)COC6H5
Quantity 100 g
Merck Index 15, 1095
Solubility Information Solubility in water: sparingly soluble
Formula Weight 212.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzoin, 98%
Show More Show Less
EINECSNumber 204-331-3
RTECSNumber DI1590000
TSCA TSCA

RUO – Research Use Only