missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, 99.5%, for analysis

Catalog No. ac221800050
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221800050 5 kg Plastic drum
AC221802500 250 g Plastic bottle
AC221800010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221800050 Supplier Thermo Scientific Chemicals Supplier No. 221800050
Only null left

CAS: 65-85-0 | C7H6O2 | 122.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00002398
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 121.0°C to 123.0°C
Boiling Point 249.0°C
CAS Min % 99.4
Color White
CAS Max % 100.0
Assay Percent Range 99.5%
Linear Formula C6H5CO2H
Beilstein 09, 92
Fieser 01,49; 13,312
Merck Index 15, 1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol, benzene, ether, chloroform,, ccl4, carbon disulfide and turpentine
Formula Weight 122.12
Percent Purity 99.5%
Grade Analytical
Density 121g/mL
Quantity 5 kg
Flash Point 121°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Fine Crystalline Powder
Chemical Name or Material Benzoic acid, For analysis
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only