missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzohydroxamic Acid 98.0+%, TCI America™
$67.11 - $266.56
Chemical Identifiers
| CAS | 495-18-1 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.138 |
| MDL Number | MFCD00002109 |
| InChI Key | VDEUYMSGMPQMIK-UHFFFAOYSA-N |
| Synonym | benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid |
| PubChem CID | 10313 |
| IUPAC Name | N-hydroxybenzamide |
| SMILES | C1=CC=C(C=C1)C(=O)NO |
Chemical Identifiers
| 495-18-1 | |
| 137.138 | |
| VDEUYMSGMPQMIK-UHFFFAOYSA-N | |
| 10313 | |
| C1=CC=C(C=C1)C(=O)NO |
| C7H7NO2 | |
| MFCD00002109 | |
| benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid | |
| N-hydroxybenzamide |
Specifications
| 495-18-1 | |
| White-Yellow | |
| MFCD00002109 | |
| benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid | |
| C1=CC=C(C=C1)C(=O)NO | |
| 137.138 | |
| 137.14 | |
| Crystalline Powder |
| 124°C | |
| C7H7NO2 | |
| 5 g | |
| VDEUYMSGMPQMIK-UHFFFAOYSA-N | |
| N-hydroxybenzamide | |
| 10313 | |
| ≥98.0% (T) | |
| Benzohydroxamic Acid |
Safety and Handling
RTECSNumber : DF9650000
TSCA : Yes