missing translation for 'onlineSavingsMsg'
Learn More

Benzohydroxamic Acid 98.0+%, TCI America™

Catalog No. B00615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00615G 5 g
B006125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B00615G Supplier TCI America Supplier No. B00615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 495-18-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00002109
InChI Key VDEUYMSGMPQMIK-UHFFFAOYSA-N
Synonym benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
PubChem CID 10313
IUPAC Name N-hydroxybenzamide
SMILES C1=CC=C(C=C1)C(=O)NO

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 137.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzohydroxamic Acid
RTECSNumber DF9650000
TSCA Yes