missing translation for 'onlineSavingsMsg'
Learn More

Benzofuroxan 98.0+%, TCI America™

Catalog No. p-7118988
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B131625G 25 g
B1316500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B131625G Supplier TCI America Supplier No. B131625G
Only null left

Chemical Identifiers

CAS 480-96-6
Molecular Formula C6H4N2O2
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005807
InChI Key OKEAMBAZBICIFP-UHFFFAOYSA-N
Synonym benzofuroxan, benzofuroxane, benzofurazan 1-oxide, benzofurazan oxide, 2,1,3-benzoxadiazole 1-oxide, 2,1,3-benzoxadiazole, 1-oxide, benzofurazan, 1-oxide, 2,1,3-benzoxadiazol-1-ium-1-olate, benzo c 1,2,5-oxadiazol-1-ol, benzofurazan n-oxide
PubChem CID 68075
IUPAC Name 3-oxido-2,1,3-benzoxadiazol-3-ium
SMILES C1=CC2=NO[N+](=C2C=C1)[O-]

Specifications

Melting Point 69°C
Color Yellow
Quantity 25 g
Formula Weight 136.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzofuroxan
EINECSNumber (5)-0599
TSCA Yes