missing translation for 'onlineSavingsMsg'
Learn More

Benzofurazan-4-sulfonyl chloride, 97%

Catalog No. p-7036034
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54275MD 250 mg
AAH5427503 1 g
AAH5427506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54275MD Supplier Thermo Scientific Chemicals Supplier No. H54275MD
Only null left

CAS: 114322-14-4 | C6H3ClN2O3S | 218.611 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114322-14-4
Molecular Formula C6H3ClN2O3S
Molecular Weight (g/mol) 218.611
MDL Number MFCD00068072
InChI Key SIYWVPMYOGMRDW-UHFFFAOYSA-N
Synonym benzofurazan-4-sulfonyl chloride, benzofurazan-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, benzo 1,2,5 oxadiazole-4-sulfonyl chloride, benzo c 1,2,5 oxadiazole-4-sulfonyl chloride, 2,1,3-benzoxadiazole-4-sulphonyl chloride, acmc-20echy, pubchem5461, 4-chlorosulphonylbenzofurazan, benzofurazan4-sulfonyl chloride
PubChem CID 2735446
IUPAC Name 2,1,3-benzoxadiazole-4-sulfonyl chloride
SMILES C1=CC2=NON=C2C(=C1)S(=O)(=O)Cl

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 218.61
Percent Purity 97%
Chemical Name or Material Benzofurazan-4-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only