missing translation for 'onlineSavingsMsg'
Learn More

Benzofuran-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B45221G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45221G Supplier TCI America Supplier No. B45221G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 317830-83-4
Molecular Formula C8H7BO3
MDL Number MFCD06801687
InChI Key DFUGYZQSDFQVPU-UHFFFAOYSA-N
PubChem CID 23438904

Specifications

Quantity 1 g
Synonym benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid
SMILES OB(O)C1=COC2=CC=CC=C12
IUPAC Name (1-benzofuran-3-yl)boronic acid
Molecular Weight (g/mol) 161.95
Formula Weight 161.95
Physical Form Crystalline Powder
Chemical Name or Material Benzofuran-3-boronic Acid (contains varying amounts of Anhydride)