missing translation for 'onlineSavingsMsg'
Learn More

Benzofuran-2-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B29781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29781G Supplier TCI America Supplier No. B29781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98437-24-2
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD00236019
InChI Key PKRRNTJIHGOMRC-UHFFFAOYSA-N
Synonym benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PubChem CID 2776266
IUPAC Name (1-benzofuran-2-yl)boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2O1

Specifications

Melting Point 135°C
Quantity 1 g
Formula Weight 161.95
Physical Form Crystalline Powder
Chemical Name or Material Benzofuran-2-boronic Acid (contains varying amounts of Anhydride)