missing translation for 'onlineSavingsMsg'
Learn More

Benzo[h]quinoline 99.0+%, TCI America™

Catalog No. p-7118344
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00885G 5 g
B008825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B00885G Supplier TCI America Supplier No. B00885G
Only null left

Chemical Identifiers

CAS 230-27-3
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00004984
InChI Key WZJYKHNJTSNBHV-UHFFFAOYSA-N
Synonym benzo h quinoline, 7,8-benzoquinoline, 4-azaphenanthrene, 1-naphthoquinoline, benzoquinoline, alpha-naphthoquinoline, alpha-benzoquinoline, 7,8-bnzoquinoline, .alpha.-benzoquinoline
PubChem CID 9191
IUPAC Name benzo[h]quinoline
SMILES C1=CC=C2C(=C1)C=CC3=C2N=CC=C3

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 338°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[h]quinoline
RTECSNumber DK1432000
TSCA Yes