missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[h]quinoline 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 50°C |
| Color | White-Yellow |
| Boiling Point | 338°C |
| Quantity | 25 g |
| Formula Weight | 179.22 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzo[h]quinoline |
Safety and Handling
| RTECSNumber | DK1432000 |
| TSCA | Yes |