missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene 97.0+%, TCI America™

Catalog No. p-7118347
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B00935G 5 g
B009325G 25 g
2 options

Catalog No. B00935G

Supplier: TCI America B00935G

Only null left

Chemical Identifiers

CAS 95-15-8
Molecular Formula C8H6S
Molecular Weight (g/mol) 134.196
MDL Number MFCD00005864
InChI Key FCEHBMOGCRZNNI-UHFFFAOYSA-N
Synonym benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
PubChem CID 7221
ChEBI CHEBI:35858
IUPAC Name 1-benzothiophene
SMILES C1=CC=C2C(=C1)C=CS2

Specifications

Boiling Point 221°C
Quantity 5 g
UN Number 3077
Formula Weight 134.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Benzo[b]thiophene