missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene-2-methanol 98.0+%, TCI America™
$106.25 - $357.90
Chemical Identifiers
| CAS | 17890-56-1 |
|---|---|
| Molecular Formula | C9H8OS |
| Molecular Weight (g/mol) | 164.222 |
| MDL Number | MFCD02682004 |
| InChI Key | SUHKSQTXXZQEBH-UHFFFAOYSA-N |
| Synonym | 2-hydroxymethyl benzo b thiophene, benzo b thiophene-2-methanol, benzo b thiophen-2-ylmethanol, benzo b thiophen-2-yl-methanol, 1-benzothiophen-2-yl methanol, 1-benzothien-2-ylmethanol, 2-hydroxymethyl benzothiophene, benzothiophen-2-methanol, hydroxymethylbenzothiophene, benzothiophene-2-methanol |
| PubChem CID | 2776335 |
| IUPAC Name | 1-benzothiophen-2-ylmethanol |
| SMILES | C1=CC=C2C(=C1)C=C(S2)CO |
Chemical Identifiers
| 17890-56-1 | |
| 164.222 | |
| SUHKSQTXXZQEBH-UHFFFAOYSA-N | |
| 2776335 | |
| C1=CC=C2C(=C1)C=C(S2)CO |
| C9H8OS | |
| MFCD02682004 | |
| 2-hydroxymethyl benzo b thiophene, benzo b thiophene-2-methanol, benzo b thiophen-2-ylmethanol, benzo b thiophen-2-yl-methanol, 1-benzothiophen-2-yl methanol, 1-benzothien-2-ylmethanol, 2-hydroxymethyl benzothiophene, benzothiophen-2-methanol, hydroxymethylbenzothiophene, benzothiophene-2-methanol | |
| 1-benzothiophen-2-ylmethanol |
Specifications
| 17890-56-1 | |
| White-Yellow | |
| MFCD02682004 | |
| 2-hydroxymethyl benzo b thiophene, benzo b thiophene-2-methanol, benzo b thiophen-2-ylmethanol, benzo b thiophen-2-yl-methanol, 1-benzothiophen-2-yl methanol, 1-benzothien-2-ylmethanol, 2-hydroxymethyl benzothiophene, benzothiophen-2-methanol, hydroxymethylbenzothiophene, benzothiophene-2-methanol | |
| C1=CC=C2C(=C1)C=C(S2)CO | |
| 164.222 | |
| 164.22 | |
| Crystalline Powder |
| 100°C | |
| C9H8OS | |
| 1 g | |
| SUHKSQTXXZQEBH-UHFFFAOYSA-N | |
| 1-benzothiophen-2-ylmethanol | |
| 2776335 | |
| ≥98.0% (GC) | |
| Benzo[b]thiophene-2-methanol |
Safety and Handling
TSCA : No