missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-methanol 98.0+%, TCI America™

Catalog No. B46285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46285G 5 g
B46281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46285G Supplier TCI America Supplier No. B46285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17890-56-1
Molecular Formula C9H8OS
Molecular Weight (g/mol) 164.222
MDL Number MFCD02682004
InChI Key SUHKSQTXXZQEBH-UHFFFAOYSA-N
Synonym 2-hydroxymethyl benzo b thiophene, benzo b thiophene-2-methanol, benzo b thiophen-2-ylmethanol, benzo b thiophen-2-yl-methanol, 1-benzothiophen-2-yl methanol, 1-benzothien-2-ylmethanol, 2-hydroxymethyl benzothiophene, benzothiophen-2-methanol, hydroxymethylbenzothiophene, benzothiophene-2-methanol
PubChem CID 2776335
IUPAC Name 1-benzothiophen-2-ylmethanol
SMILES C1=CC=C2C(=C1)C=C(S2)CO

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 164.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]thiophene-2-methanol