missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxylic hydrazide, 97%

Catalog No. AAL1135706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1135706 5 g
AAL1135703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1135706 Supplier Thermo Scientific Chemicals Supplier No. L1135706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 175135-07-6 | C9H8N2OS | 192.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175135-07-6
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.24
MDL Number MFCD00052501
InChI Key ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carbohydrazide, benzo b thiophene-2-carboxylic hydrazide, thianaphthene-2-carboxylic hydrazide, benzo b thiophene-2-carboxylicacid, hydrazide, maybridge1_008989, acmc-1c3go, #, benzo b thiophene-2-carboxylic acid hydrazide
PubChem CID 519437
IUPAC Name 1-benzothiophene-2-carbohydrazide
SMILES NNC(=O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point 184°C to 187°C
Quantity 5 g
Beilstein 153297
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-2-carboxylic hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only