Learn More
Benzo[b]thiophene-2-carboxylic acid, 98%
CAS: 6314-28-9 | C9H6O2S | 178.21 g/mol
Supplier: Thermo Scientific Chemicals A1247106
Description
Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in ethanol, acetone and chloroform. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| Benzo[b]thiophene-2-carboxylic acid | |
| 236°C to 242°C | |
| MFCD00051636 | |
| 124613 | |
| benzo b thiophene-2-carboxylic acid, thianaphthene-2-carboxylic acid, thionaphthene-2-carboxylic acid, tnca, benzothiophene-2-carboxylic acid, 2-benzo b thiophenecarboxylic acid, unii-hlr49221fq, thionapthene-2-carboxylic acid, thianaphthene-2-carboxylicacid | |
| DYSJMQABFPKAQM-UHFFFAOYSA-N | |
| 1-benzothiophene-2-carboxylic acid | |
| 95864 | |
| 98% |
| 6314-28-9 | |
| C9H6O2S | |
| 5 g | |
| 14,9345 | |
| Soluble in ethanol,acetone and chloroform. Insoluble in water. | |
| OC(=O)C1=CC2=CC=CC=C2S1 | |
| 178.21 | |
| 178.22 |
Chemical Identifiers
| 6314-28-9 | |
| 178.21 | |
| DYSJMQABFPKAQM-UHFFFAOYSA-N | |
| 95864 | |
| OC(=O)C1=CC2=CC=CC=C2S1 |
| C9H6O2S | |
| MFCD00051636 | |
| benzo b thiophene-2-carboxylic acid, thianaphthene-2-carboxylic acid, thionaphthene-2-carboxylic acid, tnca, benzothiophene-2-carboxylic acid, 2-benzo b thiophenecarboxylic acid, unii-hlr49221fq, thionapthene-2-carboxylic acid, thianaphthene-2-carboxylicacid | |
| 1-benzothiophene-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only