missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxylic acid, 98%

Catalog No. AAA1247103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1247103 1 g
AAA1247106 5 g
AAA1247114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1247103 Supplier Thermo Scientific Chemicals Supplier No. A1247103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 6314-28-9 | C9H6O2S | 178.21 g/mol

Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in ethanol, acetone and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6314-28-9
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00051636
InChI Key DYSJMQABFPKAQM-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxylic acid, thianaphthene-2-carboxylic acid, thionaphthene-2-carboxylic acid, tnca, benzothiophene-2-carboxylic acid, 2-benzo b thiophenecarboxylic acid, unii-hlr49221fq, thionapthene-2-carboxylic acid, thianaphthene-2-carboxylicacid
PubChem CID 95864
IUPAC Name 1-benzothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point 236°C to 242°C
Quantity 1 g
Beilstein 124613
Merck Index 14,9345
Solubility Information Soluble in ethanol,acetone and chloroform. Insoluble in water.
Formula Weight 178.22
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only