missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. B37775G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37775G Supplier TCI America Supplier No. B37775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3541-37-5
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD01075041
InChI Key NXSVNPSWARVMAY-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
PubChem CID 736500
IUPAC Name 1-benzothiophene-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2S1

Specifications

Melting Point 35°C
Boiling Point 136°C
Quantity 5 g
Formula Weight 162.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Benzo[b]thiophene-2-carboxaldehyde