missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxaldehyde, 97%

Catalog No. aal1934503
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL1934503 1 g
AAL1934506 5 g
2 options

Catalog No. AAL1934503

Supplier: Thermo Scientific Chemicals L1934503

Only null left

CAS: 3541-37-5 | C9H6OS | 162.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3541-37-5
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD01075041
InChI Key NXSVNPSWARVMAY-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
PubChem CID 736500
IUPAC Name 1-benzothiophene-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2S1

Specifications

Melting Point 30°C to 32°C
Boiling Point 135°C to 136°C (8 mmHg)
Quantity 1 g
Beilstein 114170
Sensitivity Air Sensitive
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-2-carboxaldehyde
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only