missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carbohydrazide, 97%, Thermo Scientific™

Catalog No. BTB04184EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB04184EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB04184EA Supplier Thermo Scientific Chemicals Supplier No. BTB04184EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175135-07-6
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.24
MDL Number MFCD00052501
InChI Key ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carbohydrazide, benzo b thiophene-2-carboxylic hydrazide, thianaphthene-2-carboxylic hydrazide, benzo b thiophene-2-carboxylicacid, hydrazide, maybridge1_008989, acmc-1c3go, #, benzo b thiophene-2-carboxylic acid hydrazide
PubChem CID 519437
IUPAC Name 1-benzothiophene-2-carbohydrazide
SMILES NNC(=O)C1=CC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-2-carbohydrazide
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.