missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene 1,1-dioxide, 98%

Catalog No. AAA1406306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1406306 5 g
AAA1406314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1406306 Supplier Thermo Scientific Chemicals Supplier No. A1406306

CAS: 825-44-5 | C8H6O2S | 166.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 825-44-5
Molecular Formula C8H6O2S
Molecular Weight (g/mol) 166.19
MDL Number MFCD00052178
InChI Key FRJNKYGTHPUSJR-UHFFFAOYSA-N
Synonym benzo b thiophene 1,1-dioxide, benzothiophene 1,1-dioxide, thianaphthene 1,1-dioxide, benzo b thiophene, 1,1-dioxide, thianaphthene-1,1-dioxide, benzo b thiophene s,s-dioxide, benzo b thiole-1,1-dione, benzothiophene sulfone, maybridge1_005454
PubChem CID 69997
IUPAC Name 1-benzothiophene 1,1-dioxide
SMILES O=S1(=O)C=CC2=CC=CC=C12

Specifications

Melting Point 137°C to 138°C
Quantity 5 g
Beilstein 116442
Formula Weight 166.2
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene 1,1-dioxide
EINECSNumber 212-544-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only