missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-7-sulfonyl chloride, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC78203DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC78203DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

1191030-88-2
216.635
benzofuran-7-sulfonyl chloride, 7-benzofuransulfonyl chloride, benzo 2,3-b furan-7-ylchlorosulfone
1-benzofuran-7-sulfonyl chloride
C8H5ClO3S
MEOOVUNNRYYGCV-UHFFFAOYSA-N
49762852
C1=CC2=C(C(=C1)S(=O)(=O)Cl)OC=C2

Specifications

Benzo[b]furan-7-sulfonyl chloride
Amber Glass Bottle
1g
MEOOVUNNRYYGCV-UHFFFAOYSA-N
1-benzofuran-7-sulfonyl chloride
49762852
97%
1191030-88-2
C8H5ClO3S
benzofuran-7-sulfonyl chloride, 7-benzofuransulfonyl chloride, benzo 2,3-b furan-7-ylchlorosulfone
C1=CC2=C(C(=C1)S(=O)(=O)Cl)OC=C2
216.635
216.64
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents