missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-boronic acid, 98%

CAS: 317830-83-4 | C8H7BO3 | 161.95 g/mol

$138.74 - $1069.78

Chemical Identifiers

CAS 317830-83-4
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD06801687
InChI Key DFUGYZQSDFQVPU-UHFFFAOYSA-N
Synonym benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid
PubChem CID 23438904
SMILES OB(O)C1=COC2=CC=CC=C12
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64389MD
View Documents
Thermo Scientific Chemicals
H64389MD
250 mg
Each for $138.74
Only null left
 
AAH6438903
View Documents
Thermo Scientific Chemicals
H6438903
1 g
Each for $485.47
Only null left
 
AAH6438906
View Documents
Thermo Scientific Chemicals
H6438906
5 g
Each for $1,069.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

317830-83-4
161.95
DFUGYZQSDFQVPU-UHFFFAOYSA-N
23438904
C8H7BO3
MFCD06801687
benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid
OB(O)C1=COC2=CC=CC=C12

Specifications

317830-83-4
MFCD06801687
9479377
DFUGYZQSDFQVPU-UHFFFAOYSA-N
(1-benzofuran-3-yl)boronic acid
23438904
98%
Benzo[b]furan-3-boronic acid
C8H7BO3
250 mg
benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid
OB(O)C1=COC2=CC=CC=C12
161.95
161.95
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only