missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-acetonitrile, 99%

Catalog No. p-7047642
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19550MD 250 mg
AAL1955003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19550MD Supplier Thermo Scientific Chemicals Supplier No. L19550MD

CAS: 52407-43-9 | C10H7NO | 157.172 g/mol

Benzo[b]furan-3-acetonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan-3-acetonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 52407-43-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD02183555
InChI Key BJAJKVZABJZXDC-UHFFFAOYSA-N
Synonym 1-benzofuran-3-ylacetonitrile, 3-benzo b furylacetonitrile, benzofuran-3-acetonitrile, 2-benzofuran-3-yl acetonitrile, benzo b furan-3-acetonitrile, 2-3-benzofuranyl acetonitrile, 2-1-benzofuran-3-yl acetonitrile, 2-benzo b furan-3-ylethanenitrile, pubchem7006, 3-benzofuranylacetonitrile
PubChem CID 2769401
IUPAC Name 2-(1-benzofuran-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CO2)CC#N

Specifications

Melting Point 38°C to 39°C
Boiling Point 120°C to 122°C (4 mmHg)
Quantity 250 mg
Beilstein 1282785
Solubility Information Insoluble in water.
Formula Weight 157.17
Percent Purity 99%
Chemical Name or Material Benzo[b]furan-3-acetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only