missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]furan-2-carboxaldehyde, 99%
CAS: 4265-16-1 | C9H6O2 | 146.145 g/mol
$204.78 - $762.62
Chemical Identifiers
| CAS | 4265-16-1 |
|---|---|
| Molecular Formula | C9H6O2 |
| Molecular Weight (g/mol) | 146.145 |
| MDL Number | MFCD00015463 |
| InChI Key | ADDZHRRCUWNSCS-UHFFFAOYSA-N |
| Synonym | benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural |
| PubChem CID | 61341 |
| IUPAC Name | 1-benzofuran-2-carbaldehyde |
| SMILES | C1=CC=C2C(=C1)C=C(O2)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0245604
|
Thermo Scientific Chemicals
L0245604 |
2 g |
Each for $204.78
|
|
|||||
|
AAL0245609
|
Thermo Scientific Chemicals
L0245609 |
10 g |
Each for $762.62
|
|
|||||
Description
Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Chemical Identifiers
| 4265-16-1 | |
| 146.145 | |
| ADDZHRRCUWNSCS-UHFFFAOYSA-N | |
| 61341 | |
| C1=CC=C2C(=C1)C=C(O2)C=O |
| C9H6O2 | |
| MFCD00015463 | |
| benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural | |
| 1-benzofuran-2-carbaldehyde |
Specifications
| 4265-16-1 | |
| C9H6O2 | |
| MFCD00015463 | |
| 2910 | |
| benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural | |
| ADDZHRRCUWNSCS-UHFFFAOYSA-N | |
| 1-benzofuran-2-carbaldehyde | |
| 61341 | |
| 99% |
| 92°C to 97°C (1 mmHg) | |
| 1.632 | |
| 2 g | |
| Air sensitive | |
| Not miscible or difficult to mix with water. | |
| C1=CC=C2C(=C1)C=C(O2)C=O | |
| 146.145 | |
| 146.15 | |
| Benzo[b]furan-2-carboxaldehyde |
Safety and Handling
EINECSNumber : 224-248-6
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only