missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxaldehyde, 99%

Catalog No. AAL0245604
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0245604 2 g
AAL0245609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0245604 Supplier Thermo Scientific Chemicals Supplier No. L0245604
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4265-16-1 | C9H6O2 | 146.145 g/mol

Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.

Chemical Identifiers

CAS 4265-16-1
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD00015463
InChI Key ADDZHRRCUWNSCS-UHFFFAOYSA-N
Synonym benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
PubChem CID 61341
IUPAC Name 1-benzofuran-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(O2)C=O

Specifications

Boiling Point 92°C to 97°C (1 mmHg)
Refractive Index 1.632
Quantity 2 g
Beilstein 2910
Sensitivity Air sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 146.15
Percent Purity 99%
Chemical Name or Material Benzo[b]furan-2-carboxaldehyde
EINECSNumber 224-248-6
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only