missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxaldehyde, 96%, Thermo Scientific™

$121.40

Chemical Identifiers

CAS 4265-16-1
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.14
MDL Number MFCD00015463
InChI Key ADDZHRRCUWNSCS-UHFFFAOYSA-N
Synonym benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
PubChem CID 61341
IUPAC Name 1-benzofuran-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(O2)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC330140010
View Documents Promotion Details
N/A 1g
Each for $121.40
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4265-16-1
146.14
ADDZHRRCUWNSCS-UHFFFAOYSA-N
61341
C1=CC=C2C(=C1)C=C(O2)C=O
C9H6O2
MFCD00015463
benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
1-benzofuran-2-carbaldehyde

Specifications

4265-16-1
100.0
>110°C
95% min. (GC)
C9H6O2
MFCD00015463
benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
ADDZHRRCUWNSCS-UHFFFAOYSA-N
1-benzofuran-2-carbaldehyde
61341
96%
95.0
1.2060g/mL
Authentic
Glass bottle
1.6300 to 1.6330
1.206
Solubility in water: .. Other solubilities: acetone,ethanol,halogenated solvents
C1=CC=C2C(=C1)C=C(O2)C=O
146.14
146.14
Benzo[b]furan-2-carboxaldehyde, 96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 224-248-6

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only