missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]furan-2-boronic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 98437-24-2 |
|---|---|
| Molecular Formula | C8H7BO3 |
| Molecular Weight (g/mol) | 161.95 |
| MDL Number | MFCD00236019 |
| InChI Key | PKRRNTJIHGOMRC-UHFFFAOYSA-N |
| Synonym | benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid |
| PubChem CID | 2776266 |
| IUPAC Name | 1-benzofuran-2-ylboronic acid |
| SMILES | B(C1=CC2=CC=CC=C2O1)(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC378730010
|
Thermo Scientific Chemicals
378730010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC378730050
|
Thermo Scientific Chemicals
378730050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC378730250
|
Thermo Scientific Chemicals
378730250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 98437-24-2 | |
| 161.95 | |
| PKRRNTJIHGOMRC-UHFFFAOYSA-N | |
| 2776266 | |
| B(C1=CC2=CC=CC=C2O1)(O)O |
| C8H7BO3 | |
| MFCD00236019 | |
| benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid | |
| 1-benzofuran-2-ylboronic acid |
Specifications
| 98437-24-2 | |
| 100.0 | |
| 96% min. (HPLC) | |
| C8H7BO3 | |
| 1g | |
| Solubility in water: ca 2.5% | |
| B(C1=CC2=CC=CC=C2O1)(O)O | |
| 161.95 | |
| 161.95 | |
| Benzo[b]furan-2-boronic acid |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00236019 | |
| benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid | |
| PKRRNTJIHGOMRC-UHFFFAOYSA-N | |
| 1-benzofuran-2-ylboronic acid | |
| 2776266 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
RUO â Research Use Only