missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo-12-crown 4-Ether 98.0+%, TCI America™
$229.29 - $747.49
Chemical Identifiers
| CAS | 14174-08-4 |
|---|---|
| Molecular Formula | C12H16O4 |
| Molecular Weight (g/mol) | 224.26 |
| MDL Number | MFCD00059934 |
| InChI Key | OAJNZFCPJVBYHB-UHFFFAOYSA-N |
| Synonym | 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene |
| PubChem CID | 586075 |
| IUPAC Name | 2,3,5,6,8,9-hexahydro-1,4,7,10-benzotetraoxacyclododecine |
| SMILES | C1COCCOC2=CC=CC=C2OCCO1 |
Chemical Identifiers
| 14174-08-4 | |
| 224.26 | |
| OAJNZFCPJVBYHB-UHFFFAOYSA-N | |
| 586075 | |
| C1COCCOC2=CC=CC=C2OCCO1 |
| C12H16O4 | |
| MFCD00059934 | |
| 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene | |
| 2,3,5,6,8,9-hexahydro-1,4,7,10-benzotetraoxacyclododecine |
Specifications
| 14174-08-4 | |
| White | |
| MFCD00059934 | |
| 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene | |
| C1COCCOC2=CC=CC=C2OCCO1 | |
| 224.26 | |
| 224.26 | |
| Crystalline Powder |
| 46°C | |
| C12H16O4 | |
| 1 g | |
| OAJNZFCPJVBYHB-UHFFFAOYSA-N | |
| 2,3,5,6,8,9-hexahydro-1,4,7,10-benzotetraoxacyclododecine | |
| 586075 | |
| ≥98.0% (GC) | |
| Benzo-12-crown 4-Ether |
Safety and Handling
TSCA : No