Learn More
ACROBiosystems Inc ACROBiosystems BenzNuclease DNA and RNA Nuclease (Ultra Pure,Protease Free)
Catalog No. 502106778
Encompass
Recombinant Protein;10KU;BenzNuclease-DNA-RNA-Nuclease (BEE-N3116) is expressed from E.coli cells.;BenzNuclease is a recombinant form of Serratia macescens extracellular endonuclease produced in Escherichia coli cells using a proprietary process at ACRObiosystems. BenzNuclease is a homodimer with monomer molecular masses about 30 kDa. Two disulfide bonds found in the nuclease are crucial to its activity and stability. The enzyme is a non-specific nuclease with high specific activity, which degrades both single- and double-stranded nucleic acids in any form ( single stranded, double stranded, linear, circular and supercoiled ). It hydrolyzes internal phosphodiester bonds present between the nucleotides to 5'-phosphorylated oligonucleotides of 3-8 bases in length.
Catalog No. 502106778 Supplier ACROBiosystems Inc Supplier No. BEEN311610KU
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;10KU;BenzNuclease-DNA-RNA-Nuclease (BEE-N3116) is expressed from E.coli cells.;BenzNuclease is a recombinant form of Serratia macescens extracellular endonuclease produced in Escherichia coli cells using a proprietary process at ACRObiosystems. BenzNuclease is a homodimer with monomer molecular masses about 30 kDa. Two disulfide bonds found in the nuclease are crucial to its activity and stability. The enzyme is a non-specific nuclease with high specific activity, which degrades both single- and double-stranded nucleic acids in any form ( single stranded, double stranded, linear, circular and supercoiled ). It hydrolyzes internal phosphodiester bonds present between the nucleotides to 5'-phosphorylated oligonucleotides of 3-8 bases in length.