missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzimidazole 98.0+%, TCI America™
$35.41 - $264.70
Chemical Identifiers
| CAS | 51-17-2 |
|---|---|
| Molecular Formula | C7H6N2 |
| Molecular Weight (g/mol) | 118.14 |
| MDL Number | MFCD00005585 |
| InChI Key | HYZJCKYKOHLVJF-UHFFFAOYSA-N |
| Synonym | benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene |
| PubChem CID | 5798 |
| ChEBI | CHEBI:41275 |
| IUPAC Name | 1H-1,3-benzodiazole |
| SMILES | N1C=NC2=CC=CC=C12 |
Chemical Identifiers
| 51-17-2 | |
| 118.14 | |
| HYZJCKYKOHLVJF-UHFFFAOYSA-N | |
| 5798 | |
| 1H-1,3-benzodiazole |
| C7H6N2 | |
| MFCD00005585 | |
| benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene | |
| CHEBI:41275 | |
| N1C=NC2=CC=CC=C12 |
Specifications
| 51-17-2 | |
| C7H6N2 | |
| 25 g | |
| HYZJCKYKOHLVJF-UHFFFAOYSA-N | |
| 1H-1,3-benzodiazole | |
| 5798 | |
| 118.14 | |
| Crystalline Powder |
| 173°C | |
| MFCD00005585 | |
| benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene | |
| N1C=NC2=CC=CC=C12 | |
| 118.14 | |
| CHEBI:41275 | |
| ≥98.0% (GC,T) | |
| Benzimidazole |
Safety and Handling
EINECSNumber : (5)-3548
RTECSNumber : DD5425000
TSCA : Yes