missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole 98.0+%, TCI America™

Catalog No. B005425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B005425G 25 g
B0054100G 100 g
B0054500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B005425G Supplier TCI America Supplier No. B005425G

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
IUPAC Name 1H-1,3-benzodiazole
SMILES N1C=NC2=CC=CC=C12

Specifications

Melting Point 173°C
Quantity 25 g
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzimidazole
EINECSNumber (5)-3548
RTECSNumber DD5425000
TSCA Yes