missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole, 99%

CAS: 51-17-2 | C7H6N2 | 118.14 g/mol

$190.66 - $631.97

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
SMILES N1C=NC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1276330
View Documents
Thermo Scientific Chemicals
A1276330
250 g N/A
Only null left
 
AAA127630B
View Documents
Thermo Scientific Chemicals
A127630B
1000 g N/A
Only null left
 
Description

Description

Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

51-17-2
118.14
HYZJCKYKOHLVJF-UHFFFAOYSA-N
5798
N1C=NC2=CC=CC=C12
C7H6N2
MFCD00005585
benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
CHEBI:41275

Specifications

51-17-2
360°C
MFCD00005585
109682
benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
HYZJCKYKOHLVJF-UHFFFAOYSA-N
1H-1,3-benzodiazole
5798
118.14
Benzimidazole
170°C to 174°C
C7H6N2
250 g
14,1081
Sparingly soluble in water.
N1C=NC2=CC=CC=C12
118.14
CHEBI:41275
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-081-4

RTECSNumber : DD5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only