missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole-5-carboxylic acid, 98%

CAS: 15788-16-6 | C8H6N2O2 | 162.15 g/mol

$150.93 - $2420.23

Chemical Identifiers

CAS 15788-16-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00011555
InChI Key COYPLDIXZODDDL-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid
PubChem CID 459456
SMILES OC(=O)C1=CC=C2N=CNC2=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1598009
View Documents
Thermo Scientific Chemicals
A1598009
10 g
Each for $150.93
Only null left
 
AAA1598014
View Documents
Thermo Scientific Chemicals
A1598014
25 g
Each for $328.34
Only null left
 
AAA1598018
View Documents
Thermo Scientific Chemicals
A1598018
50 g
Each for $598.44
Only null left
 
AAA1598030
View Documents
Thermo Scientific Chemicals
A1598030
250 g
Each for $2,420.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15788-16-6
162.15
COYPLDIXZODDDL-UHFFFAOYSA-N
459456
C8H6N2O2
MFCD00011555
1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid
OC(=O)C1=CC=C2N=CNC2=C1

Specifications

15788-16-6
C8H6N2O2
10 g
1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid
OC(=O)C1=CC=C2N=CNC2=C1
162.15
162.15
Benzimidazole-5-carboxylic acid
>300°C
MFCD00011555
5686
COYPLDIXZODDDL-UHFFFAOYSA-N
1H-1,3-benzodiazole-6-carboxylic acid
459456
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only