missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole-5,6-dicarboxylic acid, 97%

Catalog No. AAL1475406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1475406 5 g
AAL1475403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1475406 Supplier Thermo Scientific Chemicals Supplier No. L1475406

CAS: 10351-75-4 | C9H6N2O4 | 206.16 g/mol

Benzimidazole-5,6-dicarboxylic acid is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole-5,6-dicarboxylic acid is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10351-75-4
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.16
MDL Number MFCD03093058
InChI Key PIPQOFRJDBZPFR-UHFFFAOYSA-N
Synonym benzimidazole-5,6-dicarboxylic acid, 1h-benzo d imidazole-5,6-dicarboxylic acid, 1h-1,3-benzodiazole-5,6-dicarboxylic acid, 1h-benzoimidazole-5,6-dicarboxylic, 1h-benzoimidazole-5,6-dicarboxylic acid, acmc-1bsyc, 5,6-benzimidazoledicarboxylic acid, benzo d imidazole-5,6-dicarboxylic acid
PubChem CID 7023591
IUPAC Name 1H-benzimidazole-5,6-dicarboxylic acid
SMILES OC(=O)C1=C(C=C2N=CNC2=C1)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Quantity 5 g
Beilstein 528170
Solubility Information Insoluble in water.
Formula Weight 206.16
Percent Purity 97%
Chemical Name or Material Benzimidazole-5,6-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only