missing translation for 'onlineSavingsMsg'
Learn More

Benzil 99.0+%, TCI America™

Catalog No. p-7118322
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B005025G 25 g
B0050500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B005025G Supplier TCI America Supplier No. B005025G
Only null left

Chemical Identifiers

CAS 134-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00003080
InChI Key WURBFLDFSFBTLW-UHFFFAOYSA-N
Synonym benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
PubChem CID 8651
ChEBI CHEBI:51507
IUPAC Name diphenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C
Color Yellow
Boiling Point 348°C
Quantity 25 g
Formula Weight 210.23
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzil
EINECSNumber (4)-0078
RTECSNumber DD1925000
TSCA Yes