missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrazide, 98%

Catalog No. AAA1226836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1226836 500 g
AAA1226814 25 g
AAA1226822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1226836 Supplier Thermo Scientific Chemicals Supplier No. A1226836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 613-94-5 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-94-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007596
InChI Key WARCRYXKINZHGQ-UHFFFAOYSA-N
Synonym benzoylhydrazine, benzhydrazide, benzoic hydrazide, benzoyl hydrazine, benzoic acid, hydrazide, benzoic acid hydrazide, benzohydrazine, benzoylhydrazide, benzoyl hydrazide, hydrazine, benzoyl
PubChem CID 11955
ChEBI CHEBI:38454
IUPAC Name benzohydrazide
SMILES NNC(=O)C1=CC=CC=C1

Specifications

Melting Point 113°C to 117°C
Quantity 500 g
Beilstein 471797
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material Benzhydrazide
Hazard Category H302-H315-H319-H335-H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 210-363-9
RTECSNumber DH1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only