missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic Anhydride 96.0+%, TCI America™

Catalog No. B19315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19315G 5 g
B193125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19315G Supplier TCI America Supplier No. B19315G
Only null left

Chemical Identifiers

CAS 512-35-6
Molecular Formula C12H10O5S2
Molecular Weight (g/mol) 298.327
MDL Number MFCD00054601
InChI Key MLWPJXZKQOPTKZ-UHFFFAOYSA-N
PubChem CID 11335509
IUPAC Name benzenesulfonyl benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=CC=C2

Specifications

Quantity 5 g
UN Number 3261
Formula Weight 298.33
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonic Anhydride