missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, tech. ca 75% w/w aq. soln.

Catalog No. AAL19077AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19077AK 250 mL
AAL19077AU 1000 mL
AAL190770J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19077AK Supplier Thermo Scientific Chemicals Supplier No. L19077AK
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-11-3 | C6H6O3S | 158.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-11-3
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.171
MDL Number MFCD00011689
InChI Key SRSXLGNVWSONIS-UHFFFAOYSA-N
Synonym benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
PubChem CID 7371
ChEBI CHEBI:64455
IUPAC Name benzenesulfonic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Density 1.3
Flash Point >110°C (230°F)
Quantity 250 mL
UN Number UN2586
Beilstein 742513
Merck Index 14,1070
Formula Weight 158.18
Grade Technical
Concentration or Composition (by Analyte or Components) ≈75% w/w aq. soln.
Chemical Name or Material Benzenesulfonic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID
EINECSNumber 202-638-7
RTECSNumber DB4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only