missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid isopropyl ester, 95%

Catalog No. p-7055840
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6452303 1 g
AAH6452306 5 g
AAH6452314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6452303 Supplier Thermo Scientific Chemicals Supplier No. H6452303

CAS: 6214-18-2 | C9H12O3S | 200.252 g/mol

A sulfonate ester that acts as a potential effects in drug substances impurity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 6214-18-2
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.252
MDL Number MFCD04038771
InChI Key YQZZXXKFKTWDPY-UHFFFAOYSA-N
PubChem CID 12441430
IUPAC Name propan-2-yl benzenesulfonate
SMILES CC(C)OS(=O)(=O)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Refractive Index 1.502
Quantity 1 g
Beilstein 1106314
Synonym isopropyl benzenesulfonate, isopropylbenzenesulfonate, benzenesulfonic acid, 1-methylethyl ester, benzenesulfonic acid isopropyl ester, isopropyl besylate, benzenesulfonic acid,1-methylethyl ester, benzenesulfonic acid 1-methylethyl ester
Solubility Information Soluble in chloroform and ethyl acetate.
Formula Weight 200.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Benzenesulfonic acid isopropyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DB7151080
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only