missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic Acid Anhydrous 98.0+%, TCI America™

Catalog No. B34085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34085G Supplier TCI America Supplier No. B34085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-11-3
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.171
MDL Number MFCD00011689
InChI Key SRSXLGNVWSONIS-UHFFFAOYSA-N
Synonym benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
PubChem CID 7371
ChEBI CHEBI:64455
IUPAC Name benzenesulfonic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Melting Point 59°C
Color White-Yellow
Quantity 5 g
UN Number 2585
Formula Weight 158.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonic Acid Anhydrous
EINECSNumber (3)-1928
RTECSNumber DB4200000
TSCA Yes